Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174695.3 | NP_777120.2 | 6 - 7 | 765 | 100 | |
GenBank mRNA | BC140689.1 | - | 6 - 7 | 765 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |