Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174759.2 | NP_777184.2 | 1 - 2 | 125 | 85 | |
GenBank mRNA | BC113227.1 | - | 1 - 2 | 125 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |