Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176621.3 | NP_788794.2 | 8 - 9 | 1028 | 98 | |
GenBank mRNA | AF020512.2 | - | 8 - 9 | 1028 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Bos taurus Probe spans exons |