Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_183407.2 | NP_899666.1 | 3 - 4 | 378 | 71 | |
GenBank mRNA | AY346153.2 | - | 3 - 4 | 378 | 71 | |
BC104597.1 | - | 3 - 4 | 447 | 71 | ||
BT025429.1 | - | 3 - 4 | 557 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |