Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HM13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046124.2 | NP_001039589.2 | 6 - 7 | 777 | 64 | |
GenBank mRNA | BC112454.1 | - | 6 - 7 | 759 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |