Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IHH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076870.2 | NP_001070338.2 | 2 - 3 | 388 | 64 | |
GenBank mRNA | CK967652.1 | - | 2 - 3 | 394 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Bos taurus Probe spans exons |