Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098050.1 | NP_001091519.1 | 2 - 3 | 2269 | 109 | |
GenBank mRNA | BC140537.1 | - | 2 - 3 | 2269 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |