Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPATCH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098972.2 | NP_001092442.1 | 3 - 4 | 1052 | 64 | |
GenBank mRNA | BC142442.1 | - | 3 - 4 | 1054 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |