Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFML2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100354.1 | NP_001093824.1 | 5 - 6 | 991 | 65 | |
GenBank mRNA | BC147922.1 | - | 5 - 6 | 991 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |