Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101926.2 | NP_001095396.1 | 5 - 6 | 791 | 93 | |
GenBank mRNA | BC103402.1 | - | 5 - 6 | 818 | 92 | |
BC151320.1 | - | 5 - 6 | 792 | 93 | ||
BT030699.1 | - | 5 - 6 | 436 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |