Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102489.1 | NP_001095959.1 | 9 - 10 | 885 | 113 | |
GenBank mRNA | BC140502.1 | - | 9 - 10 | 885 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |