Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCARA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102499.1 | NP_001095969.1 | 5 - 6 | 1318 | 97 | |
GenBank mRNA | BC142199.1 | - | 5 - 6 | 1318 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |