Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCDHA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102506.2 | NP_001095976.1 | 2 - 3 | 2571 | 77 | |
GenBank mRNA | BC148989.1 | - | 2 - 3 | 779 | 77 | |
BC151664.1 | - | 2 - 3 | 2572 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |