Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSPO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105621.1 | NP_001099091.1 | 5 - 6 | 1174 | 103 | |
GenBank mRNA | BC149052.1 | - | 5 - 6 | 1174 | 103 | |
BT030732.1 | - | 3 - 4 | 499 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |