Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS35L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046368.2 | NP_001039833.2 | 20 - 21 | 1710 | 88 | |
GenBank mRNA | DV907223.1 | - | 5 - 6 | 399 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |