Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS35L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046368.2 | NP_001039833.2 | 25 - 26 | 2120 | 103 | |
GenBank mRNA | DV893673.1 | - | 2 - 3 | 190 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |