Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS35L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046368.2 | NP_001039833.2 | 26 - 27 | 2235 | 91 | |
GenBank mRNA | DV893673.1 | - | 3 - 4 | 305 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |