Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099044.2 | NP_001092514.1 | 3 - 4 | 496 | 68 | |
GenBank mRNA | BC142059.1 | - | 3 - 4 | 497 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |