Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3CG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102125.2 | NP_001095595.1 | 2 - 3 | 2123 | 64 | |
GenBank mRNA | BC149468.1 | - | 2 - 3 | 2124 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |