Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL20RB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102175.2 | NP_001095645.1 | 2 - 3 | 374 | 74 | |
GenBank mRNA | BC148928.1 | - | 2 - 3 | 375 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |