Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCEANC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104992.2 | NP_001098462.1 | 4 - 5 | 654 | 132 | |
GenBank mRNA | BC142457.1 | - | 4 - 5 | 655 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |