Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EWSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109800.2 | NP_001103270.1 | 9 - 10 | 1061 | 72 | |
GenBank mRNA | BC149284.1 | - | 9 - 10 | 1025 | 72 | |
BC153844.1 | - | 9 - 10 | 1062 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |