Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMIM11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114516.2 | NP_001107988.1 | 1 - 2 | 65 | 55 | |
GenBank mRNA | BC146134.1 | - | 1 - 2 | 66 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |