Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ING1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077914.1 | NP_001071382.1 | 2 | 1045 | 78 | |
GenBank mRNA | BC120155.1 | - | 2 | 1045 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |