Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTCFL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123365.1 | NP_001116837.1 | 6 - 7 | 1143 | 75 | |
GenBank mRNA | EU527855.1 | - | 6 - 7 | 1143 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |