Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130747.1 | NP_001124219.1 | 3 - 4 | 789 | 61 | |
GenBank mRNA | BC118284.1 | - | 3 - 4 | 789 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |