Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105652.2 | NP_001099122.1 | 2 - 3 | 137 | 79 | |
GenBank mRNA | BC122769.1 | - | 2 - 3 | 139 | 79 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |