Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DKK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205544.1 | NP_001192473.1 | 2 - 3 | 517 | 71 | |
GenBank mRNA | JN966753.1 | - | 2 - 3 | 575 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Bos taurus Probe spans exons |