Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205971.1 | NP_001192900.1 | 4 - 5 | 1250 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |