Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCGB1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206224.1 | NP_001193153.1 | 2 - 3 | 252 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Bos taurus Probe spans exons |