Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPRKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192507.1 | NP_001179436.1 | 4 - 5 | 722 | 99 | |
GenBank mRNA | BC102574.1 | - | 4 - 5 | 483 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |