Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206494.1 | NP_001193423.1 | 1 - 2 | 68 | 81 | |
GenBank mRNA | BC105454.1 | - | 1 - 2 | 69 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |