Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166519.1 | NP_001159991.1 | 1 - 2 | 311 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Bos taurus Probe spans exons |