Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206633.1 | NP_001193562.1 | 7 - 8 | 2809 | 62 | |
GenBank mRNA | AF056218.1 | - | 1 - 2 | 406 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |