Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTFMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166523.3 | NP_001159995.1 | 7 - 8 | 914 | 107 | |
GenBank mRNA | AB004316.1 | - | 7 - 8 | 849 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |