Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191136.3 | NP_001178065.1 | 2 - 3 | 188 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |