Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206853.1 | NP_001193782.1 | 11 - 12 | 2120 | 65 | |
GenBank mRNA | BC149309.1 | - | 9 - 10 | 1386 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |