Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC20B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206982.1 | NP_001193911.1 | 9 - 10 | 1362 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |