Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6AP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001207027.1 | NP_001193956.1 | 7 - 8 | 843 | 78 | |
NM_001207028.1 | NP_001193957.1 | 6 - 7 | 777 | 78 | ||
GenBank mRNA | DN538471.1 | - | 1 - 2 | 151 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |