Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244183.1 | NP_001231112.1 | 1 - 2 | 180 | 93 | |
GenBank mRNA | BC118121.1 | - | 1 - 2 | 180 | 93 | |
DV801645.1 | - | 1 - 2 | 180 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |