Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLGALT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191302.2 | NP_001178231.1 | 1 - 2 | 268 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |