Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191435.1 | NP_001178364.1 | 5 - 6 | 700 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Bos taurus Probe spans exons |