Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192099.1 | NP_001179028.1 | 3 - 4 | 332 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |