Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010996.1 | NP_001010996.1 | 4 - 5 | 318 | 94 | |
GenBank mRNA | AY860499.1 | - | 4 - 5 | 318 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Bos taurus Probe spans exons |