Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076105.1 | NP_001069573.1 | 1 - 2 | 33 | 68 | |
GenBank mRNA | BC116069.1 | - | 1 - 2 | 33 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |