Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205189.1 | NP_001192118.1 | 3 - 4 | 654 | 85 | |
GenBank mRNA | BC108121.1 | - | 3 - 4 | 654 | 85 | |
CO891135.1 | - | 2 - 3 | 188 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |