Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029217.6 | NP_001024388.1 | 6 - 7 | 530 | 72 | |
NM_001029218.7 | NP_001024389.1 | 3 - 4 | 584 | 72 | ||
GenBank mRNA | AY305831.1 | - | 4 - 5 | 336 | 72 | |
AY305832.1 | - | 4 - 5 | 326 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |