Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acdh-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078388.5 | NP_510789.1 | 2 - 3 | 562 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |