Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npp-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170819.6 | NP_740802.1 | 1 - 2 | 104 | 74 | |
NR_131352.1 | 1 - 2 | 103 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |