Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sop-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306445.3 | NP_001293374.1 | 5 - 6 | 1494 | 69 | |
NM_001377813.3 | NP_001364767.1 | 10 - 11 | 2460 | 69 | ||
NM_170837.5 | NP_740821.1 | 10 - 11 | 2464 | 69 | ||
GenBank mRNA | AF308860.1 | - | 10 - 11 | 2463 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |